Search
Search

FCRLB| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002901.3 | NP_001002901.1 | 8 | 1948 | ||
| NM_001288829.1 | NP_001275758.1 | 6 | 1589 | |||
| NM_001288830.1 | NP_001275759.1 | 5 | 1568 | |||
| NM_001288831.1 | NP_001275760.1 | 6 | 1630 | |||
| NM_001288832.1 | NP_001275761.1 | 5 | 1609 | |||
| NM_001320241.1 | NP_001307170.1 | 7 | 1762 | |||
| GenBank mRNA | AK055614.1 | - | 2 | 1942 | ||
| BC038564.2 | - | 3 | 1314 | |||
| BC067080.1 | - | 2 | 1931 | |||
| DQ021957.1 | - | 8 | 1948 | |||