Search
Search

APOM| Gene Symbol: APOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256169.1 | NP_001243098.1 | 3 | 355 | ||
| NM_019101.2 | NP_061974.2 | 3 | 400 | |||
| NR_045828.1 | 3 | 362 | ||||
| XM_006715150.3 | XP_006715213.1 | 3 | 384 | |||
| GenBank mRNA | AF118393.3 | - | 3 | 360 | ||
| AF161454.1 | - | 3 | 321 | |||
| AJ245434.1 | - | 3 | 376 | |||
| BC020683.1 | - | 3 | 379 | 4733993 | ||
| BF344944.1 | - | 3 | 336 | |||
| BG564719.1 | - | 3 | 401 | |||
| CB054016.1 | - | 3 | 362 | |||
| CN428415.1 | - | 3 | 342 | |||