Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIAA1755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029864.1 | NP_001025035.1 | 14 | 4152 | ||
GenBank mRNA | AB051542.1 | - | 12 | 3610 | ||
BC011516.1 | - | 2 | 1832 | |||
BC033704.1 | - | 3 | 2067 | |||
BC047031.1 | - | 1 | 547 |