Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142648.1 | NP_001136120.1 | 4 | 320 | ||
NM_020150.4 | NP_064535.1 | 3 | 250 | |||
GenBank mRNA | AF087897.1 | - | 3 | 217 | ||
AF217959.1 | - | 3 | 217 | |||
AF261717.1 | - | 3 | 217 | |||
AF274026.1 | - | 3 | 216 | |||
AF318066.1 | - | 2 | 117 | |||
AK298591.1 | - | 2 | 83 | |||
AK312469.1 | - | 3 | 205 | |||
AL136724.1 | - | 3 | 233 | |||
AY008268.1 | - | 3 | 241 | |||
BC003658.2 | - | 3 | 215 | 3546972 | ||
BG327663.1 | - | 4 | 289 | |||
CR533492.1 | - | 2 | 117 | |||
DB216796.1 | - | 4 | 320 |