Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTBP1 SIRNA Assays ›
          Gene Symbol:
          CTBP1
          Gene Name:
          C-terminal binding protein 1
          Gene Aliases:
          BARS
          Chromosome Location:
          Chr.4: 1211440 - 1250329 on Build GRCh38
          UniGene ID:
          Hs.208597
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 2802

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTBP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012614.1 NP_001012632.1 8 1283
          NM_001328.2 NP_001319.1 7 1088
          XM_005272261.3 XP_005272318.1 7 1099
          XM_005272263.4 XP_005272320.1 8 7314
          XM_011513395.2 XP_011511697.1 8 1143
          XM_011513396.2 XP_011511698.1 8 1248
          XM_011513397.2 XP_011511699.1 7 7031
          XM_017007762.1 XP_016863251.1 8 1297
          XM_017007763.1 XP_016863252.1 8 1420
          XM_017007764.1 XP_016863253.1 8 7314
          XM_017007765.1 XP_016863254.1 8 1143
          XM_017007766.1 XP_016863255.1 8 1248
          XM_017007767.1 XP_016863256.1 8 1162
          GenBank mRNA AF091555.1 - 7 969
          BC011655.2 - 7 961 4128336
          BC053320.1 - 8 1275 6461670
          BC072021.1 - 3 282
          BC098400.1 - 7 1098 30344174
          KJ534818.1 - 7 923
          KJ535043.1 - 6 572
          KJ535050.1 - 6 768
          KJ535065.1 - 8 1118
          KJ535066.1 - 8 1118
          KJ535067.1 - 8 1118
          U37408.1 - 7 972

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019201, XM_006251225, XM_006251226, XM_006251228, XM_006251229, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001198859, NM_001198861, NM_013502, NM_001198860, AK133816, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          protein phosphorylation
          negative regulation of cell proliferation
          viral genome replication
          positive regulation of histone deacetylation
          negative regulation of histone acetylation
          negative regulation of transcription, DNA-templated
          white fat cell differentiation
          regulation of cell cycle
          oxidation-reduction process
          negative regulation of histone H4 acetylation
          negative regulation of transcription from RNA polymerase II promoter by histone modification
          RNA polymerase II transcription corepressor activity
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein C-terminus binding
          transcription factor binding
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          protein domain specific binding
          protein homodimerization activity
          NAD binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-d8jzh:80/100.66.76.85:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0