Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009622.1 | NP_001009622.1 | 7 | 950 | ||
GenBank mRNA | BC088842.1 | - | 7 | 950 | 7303357 | |
FQ215892.1 | - | 5 | 785 | |||
FQ216359.1 | - | 5 | 785 | |||
FQ216811.1 | - | 5 | 785 | |||
FQ217315.1 | - | 5 | 785 | |||
FQ217751.1 | - | 7 | 1011 | |||
FQ219007.1 | - | 5 | 785 | |||
FQ219381.1 | - | 7 | 1012 | |||
FQ220447.1 | - | 5 | 785 | |||
FQ220536.1 | - | 3 | 549 | |||
FQ224969.1 | - | 5 | 785 | |||
FQ227071.1 | - | 5 | 785 | |||
FQ233866.1 | - | 5 | 785 |