Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC690955 SIRNA Assays ›
          Gene Symbol:
          LOC690955 LOC100912456 LOC683463 LOC690948 Lilra3 Lilrb3l
          Gene Name:
          similar to paired-Ig-like receptor B
          leukocyte immunoglobulin-like receptor subfamily B member 3-like
          similar to paired-Ig-like receptor B
          similar to paired-Ig-like receptor A11
          leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3
          leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3-like
          Gene Aliases:
          -
          -
          -
          -
          -
          Lilrb3, Pir-A1, Pir-A2, Pirb
          Chromosome Location:
          Chr.1: 64074094 - 64081568 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63883682 - 63891050 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 64005794 - 64013268 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63963853 - 63971222 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63920125 - 63929449 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63759638 - 63849278 on Build Rnor_6.0
          UniGene ID:
          -
          -
          -
          -
          -
          Rn.198481
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID 282101

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC690955
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_001076302.5 XP_001076302.3 919
          XM_006228067.2 XP_006228129.1 919
          XM_006228068.2 XP_006228130.1 919
          XM_006228069.2 XP_006228131.1 725
          XM_006228070.2 XP_006228132.1 637
          XM_006228071.2 XP_006228133.1 919
          Gene Symbol: LOC100912456
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003748711.3 XP_003748759.1 5 876
          XM_006228039.2 XP_006228101.1 5 877
          XM_006228040.2 XP_006228102.1 5 875
          XM_006228042.2 XP_006228104.1 5 887
          XM_006228043.2 XP_006228105.1 4 802
          Gene Symbol: LOC683463
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003748713.3 XP_003748761.1 5 919
          XM_006228061.2 XP_006228123.1 5 919
          XM_006228062.2 XP_006228124.1 5 919
          XM_006228063.2 XP_006228125.1 5 725
          XM_006228064.2 XP_006228126.1 4 637
          XM_006228065.2 XP_006228127.1 5 919
          Gene Symbol: LOC690948
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_001076276.4 XP_001076276.1 876
          XM_006228055.2 XP_006228117.1 877
          XM_006228056.2 XP_006228118.1 875
          XM_006228058.2 XP_006228120.1 887
          XM_006228059.2 XP_006228121.1 802
          Gene Symbol: Lilra3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003748712.3 XP_003748760.1 874
          XM_006228047.2 XP_006228109.1 876
          XM_006228049.2 XP_006228111.1 875
          XM_006228050.2 XP_006228112.1 877
          XM_006228052.2 XP_006228114.1 687
          XM_006228053.2 XP_006228115.1 801
          Gene Symbol: Lilrb3l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037357.2 NP_001032434.1 801
          XM_006228032.2 XP_006228094.1 5 843
          XM_006228033.2 XP_006228095.1 5 843
          XM_006228034.2 XP_006228096.1 4 558
          GenBank mRNA AF169636.1 - 792
          AF169637.1 - 6 801
          AF169638.1 - 801

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          T cell proliferation involved in immune response
          negative regulation of cytokine secretion involved in immune response
          Fc receptor mediated inhibitory signaling pathway
          signal transduction
          response to virus
          positive regulation of gene expression
          negative regulation of serotonin secretion
          interferon-gamma production
          negative regulation of interferon-gamma production
          negative regulation of mononuclear cell proliferation
          negative regulation of interferon-beta secretion
          negative regulation of T cell proliferation
          negative regulation of tumor necrosis factor biosynthetic process
          positive regulation of apoptotic process
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of cell cycle
          negative regulation of endocytosis
          positive regulation of cytolysis
          negative regulation of natural killer cell mediated cytotoxicity
          negative regulation of alpha-beta T cell activation
          defense response to virus
          negative regulation of calcium ion transport
          cellular response to lipopolysaccharide
          interferon-gamma secretion
          dendritic cell differentiation
          negative regulation of dendritic cell apoptotic process
          negative regulation of interleukin-10 secretion
          negative regulation of interleukin-12 secretion
          negative regulation of CD8-positive, alpha-beta T cell activation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          positive regulation of gamma-delta T cell activation involved in immune response
          negative regulation of dendritic cell differentiation
          negative regulation of transforming growth factor-beta secretion
          negative regulation of osteoclast development
          protein phosphatase 1 binding
          HLA-A specific inhibitory MHC class I receptor activity
          HLA-B specific inhibitory MHC class I receptor activity
          MHC class I receptor activity
          SH2 domain binding
          MHC class I protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0