Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBX SIRNA Assays ›
Gene Symbol:
BBX
Gene Name:
BBX, HMG-box containing
Gene Aliases:
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location:
Chr.3: 107522936 - 107811329 on Build GRCh38
UniGene ID:
Hs.124366
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28336
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
283.00
Your Price
Online offer:
274.65
274.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142568.2
NP_001136040.1
4, 5
485
NM_001276286.1
NP_001263215.1
4, 5
485
NM_020235.6
NP_064620.2
4, 5
485
XM_005247642.3
XP_005247699.1
4, 5
401
XM_005247643.3
XP_005247700.1
4, 5
413
XM_005247644.3
XP_005247701.1
5, 6
576
XM_011512999.2
XP_011511301.1
4, 5
485
XM_011513000.1
XP_011511302.1
4, 5
613
XM_011513001.1
XP_011511303.1
4, 5
453
XM_011513002.2
XP_011511304.1
4, 5
485
XM_011513003.2
XP_011511305.1
4, 5
485
XM_011513004.2
XP_011511306.1
4, 5
613
XM_011513005.2
XP_011511307.1
4, 5
485
XM_011513006.2
XP_011511308.1
4, 5
485
XM_011513007.2
XP_011511309.1
4, 5
485
XM_011513010.2
XP_011511312.1
4, 5
485
XM_017006881.1
XP_016862370.1
6, 7
626
XM_017006882.1
XP_016862371.1
5, 6
576
XM_017006883.1
XP_016862372.1
4, 5
485
GenBank mRNA
AF276948.1
-
1, 2
183
AF276949.1
-
4, 5
416
AF283775.2
-
4, 5
482
AF454941.1
-
4, 5
478
AF454942.1
-
2, 3
358
AK057010.1
-
4, 5
431
AK291757.1
-
4, 5
431
AK307668.1
-
3, 4
525
AK314434.1
-
4, 5
436
AL136769.1
-
2, 3
358
BC110903.1
-
2, 3
395
BC131718.1
-
4, 5
378
40123389
DA218136.1
-
4, 5
485
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
bone development
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline