Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SF1 SIRNA Assays ›
          Gene Symbol:
          SF1
          Gene Name:
          splicing factor 1
          Gene Aliases:
          BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
          Chromosome Location:
          Chr.11: 64764604 - 64779043 on Build GRCh38
          UniGene ID:
          Hs.502829
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 284705

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178030.1 NP_001171501.1 1, 2 490
          NM_001178031.1 NP_001171502.1 1, 2 272
          NM_004630.3 NP_004621.2 1, 2 493
          NM_201995.2 NP_973724.1 1, 2 493
          NM_201997.2 NP_973726.2 1, 2 493
          NM_201998.2 NP_973727.1 1, 2 493
          NR_033649.1 1, 2 493
          NR_033650.1 1, 2 493
          XM_011545244.1 XP_011543546.1 1, 2 165
          XM_011545245.1 XP_011543547.1 1, 2 165
          XM_011545246.1 XP_011543548.1 1, 2 165
          XM_011545247.1 XP_011543549.1 1, 2 165
          XM_011545248.1 XP_011543550.1 1, 2 165
          XM_017018244.1 XP_016873733.1 1, 2 165
          XM_017018245.1 XP_016873734.1 1, 2 246
          XM_017018246.1 XP_016873735.1 1, 2 165
          XM_017018247.1 XP_016873736.1 1, 2 208
          XM_017018248.1 XP_016873737.1 1, 2 208
          XM_017018249.1 XP_016873738.1 1, 2 692
          XM_017018250.1 XP_016873739.1 1, 2 418
          XM_017018251.1 XP_016873740.1 1, 2 273
          GenBank mRNA AK293753.1 - 1, 2 272
          AK296237.1 - 1, 2 191
          AK299705.1 - 1, 2 191
          AK301803.1 - 1, 2 418
          BC008080.2 - 1, 2 189 2961666
          BC008724.2 - 1, 2 189 2961666
          BC020217.1 - 1, 2 392 4869292
          BC034451.1 - 1, 2 418
          BC038446.1 - 1, 2 513 5493676
          BX095574.1 - 1, 2 158
          D26120.1 - 1, 2 423
          D26121.1 - 1, 2 423
          DC368015.1 - 1, 2 449
          DC370979.1 - 1, 2 493
          L49345.1 - 1, 2 379
          L49380.1 - 1, 2 379
          Y08765.1 - 1, 2 83
          Y08766.1 - 1, 2 335

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          mRNA 3'-splice site recognition
          mRNA splicing, via spliceosome
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          male sex determination
          Leydig cell differentiation
          negative regulation of smooth muscle cell proliferation
          regulation of steroid biosynthetic process
          negative regulation of nucleic acid-templated transcription
          transcription corepressor activity
          RNA binding
          protein binding
          zinc ion binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0