Search
Search

Igsf8| Gene Symbol: Igsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014787.1 | NP_001014787.1 | 3, 4 | 504 | ||
| XM_006250286.2 | XP_006250348.1 | 3, 4 | 663 | |||
| XM_006250288.2 | XP_006250350.1 | 2, 3 | 605 | |||
| XM_006250289.2 | XP_006250351.1 | 2, 3 | 442 | |||
| GenBank mRNA | BC092192.1 | - | 3, 4 | 504 | 7312924 | |