Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080485.3 | NP_001073954.3 | 5 | 1438 | ||
GenBank mRNA | AK122976.1 | - | 5 | 1438 | ||
AK127133.1 | - | 1 | 1234 | |||
BC001545.1 | - | 3 | 2181 | |||
BC136634.1 | - | 2 | 2562 | 9020624 |