Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: S1pr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010102.2 | NP_034232.1 | 1 | 1414 | ||
GenBank mRNA | AJ006074.1 | - | 1 | 1414 | ||
AK075854.1 | - | 1 | 933 | |||
AK154242.1 | - | 1 | 937 | |||
BC107000.1 | - | 1 | 984 | 40045304 |
Set Membership: |
G-protein coupled receptor |