Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNAS SIRNA Assays ›
          Gene Symbol:
          Gnas
          Gene Name:
          GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus
          Gene Aliases:
          5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH
          Chromosome Location:
          Chr.2: 174283444 - 174346744 on Build GRCm38
          UniGene ID:
          Mm.125770
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 287387

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gnas
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077507.1 NP_001070975.1 2 2376
          NM_001077510.2 NP_001070978.1 2 363
          NM_010309.3 NP_034439.2 2 2376
          NM_019690.2 NP_062664.2 3 1081
          NM_022000.2 NP_068840.2 3 1081
          NM_201616.1 NP_963910.1 2 363
          NM_201618.1 NP_963912.1 2 2376
          GenBank mRNA AF107847.1 - 3 1085
          AF107848.1 - 3 1079
          AF116268.1 - 2 1305
          AF125259.1 - 2 478
          AF175305.1 - 3 1056
          AJ010163.1 - 3 1047
          AK078037.1 - 2 377
          AK085136.1 - 1 1134
          AK132636.1 - 2 464
          AK147051.1 - 2 361
          AK159563.1 - 2 364
          AK168996.1 - 2 334
          AK197380.1 - 2 33
          AY519501.1 - 2 2376
          AY519502.1 - 2 2376
          AY519504.1 - 3 2471
          AY682719.1 - 3 69
          BC013824.1 - 2 176
          BC013840.1 - 2 176
          BC034446.1 - 2 458
          BC038067.1 - 3 375
          BC061496.1 - 2 347
          BC062654.1 - 3 2518
          BC080816.1 - 2 352
          BC092055.1 - 2 167
          BC106133.1 - 2 169
          BF468141.1 - 2 355
          BY718446.1 - 2 137
          M13964.1 - 2 166
          Y00703.1 - 2 175

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001159653, NM_001024823, NM_019132, NM_021845, NM_001159656, and others.

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          tissue homeostasis
          positive regulation of protein phosphorylation
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          regulation of transcription from RNA polymerase II promoter
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          post-embryonic development
          regulation of signal transduction
          positive regulation of sodium ion transport
          positive regulation of cAMP biosynthetic process
          embryonic hindlimb morphogenesis
          multicellular organism growth
          negative regulation of renal sodium excretion
          negative regulation of multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of GTPase activity
          skin development
          positive regulation of cAMP-mediated signaling
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          negative regulation of blood pressure
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          calcium ion homeostasis
          bone development
          hair follicle placode formation
          platelet aggregation
          response to parathyroid hormone
          cellular response to prostaglandin E stimulus
          genetic imprinting
          cellular response to catecholamine stimulus
          adenylate cyclase-activating adrenergic receptor signaling pathway
          nucleotide binding
          G-protein alpha-subunit binding
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          guanyl nucleotide binding
          protein domain specific binding
          G-protein beta-subunit binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          alpha-tubulin binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dcnqv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0