Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCK2 SIRNA Assays ›
          Gene Symbol:
          NCK2
          Gene Name:
          NCK adaptor protein 2
          Gene Aliases:
          GRB4, NCKbeta
          Chromosome Location:
          Chr.2: 105744649 - 105894274 on Build GRCh38
          UniGene ID:
          Hs.529244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 288565

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001004720.2 NP_001004720.1 4 1894
          NM_001004722.3 NP_001004722.1 4 1282
          NM_003581.4 NP_003572.2 5 2004
          XM_006712797.3 XP_006712860.1 5 2660
          XM_011511991.2 XP_011510293.1 5 2098
          XM_011511992.2 XP_011510294.2 3 1226
          XM_017005103.1 XP_016860592.1 6 2262
          XM_017005104.1 XP_016860593.1 5 2274
          XM_017005105.1 XP_016860594.1 5 2011
          GenBank mRNA BC000103.2 - 4 1782 3505427
          BC007195.1 - 4 1789 3608149

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immunological synapse formation
          actin filament organization
          signal transduction
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          regulation of epidermal growth factor-activated receptor activity
          negative regulation of cell proliferation
          vesicle-mediated transport
          cell migration
          lamellipodium assembly
          positive regulation of actin filament polymerization
          negative regulation of peptidyl-serine phosphorylation
          positive regulation of translation in response to endoplasmic reticulum stress
          positive regulation of T cell proliferation
          T cell activation
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          dendritic spine development
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of PERK-mediated unfolded protein response
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          SH3/SH2 adaptor activity
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          cytoskeletal adaptor activity
          receptor signaling complex scaffold activity
          protein complex binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0