Search
Search

CRABP2| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199723.1 | NP_001186652.1 | 2 | 267 | ||
| NM_001878.3 | NP_001869.1 | 1 | 280 | |||
| XM_017000343.1 | XP_016855832.1 | 4 | 5694 | |||
| GenBank mRNA | AB593017.1 | - | 1 | 196 | ||
| AK312007.1 | - | 1 | 196 | |||
| AU126559.1 | - | 2 | 267 | |||
| BC001109.2 | - | 1 | 182 | 2988143 | ||
| BP231157.1 | - | 1 | 280 | |||
| BT019827.1 | - | 1 | 59 | |||
| CR450357.1 | - | 1 | 59 | |||
| M68867.1 | - | 1 | 157 | |||