Search
Search

PVALB| Gene Symbol: PVALB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001315532.1 | NP_001302461.1 | 3 | 267 | ||
| NM_002854.2 | NP_002845.1 | 4 | 287 | |||
| GenBank mRNA | AK311832.1 | - | 3 | 267 | ||
| AV724364.1 | - | 3 | 246 | |||
| BC069300.1 | - | 4 | 287 | 7262744 | ||
| BC096112.3 | - | 4 | 293 | 40002305 | ||
| BC096113.3 | - | 4 | 293 | 40002307 | ||
| BC096114.1 | - | 4 | 293 | 40002308 | ||
| BC096115.1 | - | 4 | 293 | 40002310 | ||
| CR456552.1 | - | 3 | 253 | |||
| DA054941.1 | - | 3 | 268 | |||
| DA392417.1 | - | 3 | 267 | |||
| X63070.1 | - | 4 | 293 | |||