Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004375.4 | NP_004366.1 | 4 | 810 | ||
NR_135677.1 | 4 | 810 | ||||
XM_011524342.2 | XP_011522644.1 | 4 | 3049 | |||
XM_017024192.1 | XP_016879681.1 | 4 | 3049 | |||
XM_017024193.1 | XP_016879682.1 | 4 | 3049 | |||
GenBank mRNA | AF044321.1 | - | 4 | 686 | ||
AK293851.1 | - | 4 | 643 | |||
BC005895.1 | - | 4 | 681 | 4282744 | ||
CR541837.1 | - | 4 | 671 | |||
U79270.1 | - | 3 | 301 |