Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALS2 SIRNA Assays ›
          Gene Symbol:
          ALS2
          Gene Name:
          ALS2, alsin Rho guanine nucleotide exchange factor
          Gene Aliases:
          ALS2CR6, ALSJ, IAHSP, PLSJ
          Chromosome Location:
          Chr.2: 201700263 - 201781172 on Build GRCh38
          UniGene ID:
          Hs.471096
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 28994

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020919.3 NP_065970.2 34 5539
          XM_006712654.2 XP_006712717.1 34 6116
          XM_006712655.3 XP_006712718.1 30 3753
          XM_011511530.2 XP_011509832.1 33 5251
          XM_017004569.1 XP_016860058.1 33 5240
          XM_017004571.1 XP_016860060.1 33 5178
          XM_017004572.1 XP_016860061.1 31 4831
          GenBank mRNA AB046783.2 - 33 5260
          AB053305.1 - 34 5306
          AF391100.1 - 34 5306

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          in utero embryonic development
          receptor recycling
          response to oxidative stress
          endosome organization
          axonogenesis
          neuromuscular junction development
          locomotory behavior
          protein localization
          cell death
          vesicle organization
          endosomal transport
          Rac protein signal transduction
          positive regulation of Rac protein signal transduction
          regulation of Rho protein signal transduction
          synaptic transmission, glutamatergic
          positive regulation of GTPase activity
          positive regulation of protein kinase activity
          neuron projection morphogenesis
          regulation of endosome size
          positive regulation of protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Ran guanyl-nucleotide exchange factor activity
          protein binding
          Rab guanyl-nucleotide exchange factor activity
          Rab GTPase binding
          Rac guanyl-nucleotide exchange factor activity
          protein homodimerization activity
          protein serine/threonine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-w7qhp:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0