Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190791.1 | NP_001177720.1 | 6 | 2574 | ||
NM_020933.4 | NP_065984.3 | 7 | 2670 | |||
NR_102435.1 | 7 | 2816 | ||||
GenBank mRNA | AB046808.1 | - | 6 | 2447 | ||
AF148135.1 | - | 6 | 2557 | |||
AF275255.1 | - | 7 | 2653 | |||
AK056205.1 | - | 1 | 1534 | |||
AK056570.1 | - | 7 | 2653 | |||
AL834126.2 | - | 5 | 3745 | |||
BC078154.1 | - | 7 | 2640 | 6060418 |