Search
Search

SLC5A6| Gene Symbol: SLC5A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021095.2 | NP_066918.2 | 7 | 1094 | ||
| NR_028323.1 | 7 | 1094 | ||||
| XM_006712128.1 | XP_006712191.1 | 7 | 889 | |||
| XM_006712129.1 | XP_006712192.1 | 7 | 937 | |||
| XM_006712130.1 | XP_006712193.1 | 7 | 928 | |||
| XM_011533146.2 | XP_011531448.1 | 5 | 716 | |||
| XM_017005216.1 | XP_016860705.1 | 4 | 664 | |||
| GenBank mRNA | AF069307.1 | - | 7 | 990 | ||
| AF081571.1 | - | 6 | 806 | |||
| AK027855.1 | - | 7 | 935 | |||
| AK314545.1 | - | 8 | 921 | |||
| BC012806.2 | - | 5 | 656 | 3507776 | ||
| BC015631.2 | - | 6 | 807 | 3628126 | ||