Search Thermo Fisher Scientific
Gene Symbol: KLK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019598.2 | NP_062544.1 | 5 | 614 | ||
NM_145894.1 | NP_665901.1 | 5 | 614 | |||
NM_145895.1 | NP_665902.1 | 4 | 354 | |||
XM_005258950.3 | XP_005259007.1 | 4 | 1262 | |||
XM_005258951.3 | XP_005259008.1 | 4 | 1099 | |||
GenBank mRNA | AY358524.1 | - | 4 | 617 | ||
BC035385.1 | - | 4 | 841 | 4825490 | ||
BC136341.1 | - | 4 | 313 | 9020328 | ||
BC143988.1 | - | 4 | 473 | |||
BC143990.1 | - | 4 | 313 | |||
BC143991.1 | - | 5 | 669 | |||
BX648580.1 | - | 2 | 265 |