Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDA2R SIRNA Assays ›
          Gene Symbol:
          EDA2R
          Gene Name:
          ectodysplasin A2 receptor
          Gene Aliases:
          EDA-A2R, EDAA2R, TNFRSF27, XEDAR
          Chromosome Location:
          Chr.X: 66594384 - 66639303 on Build GRCh38
          UniGene ID:
          Hs.302017
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 29336

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EDA2R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199687.2 NP_001186616.1 2, 3 191
          NM_001242310.1 NP_001229239.1 1, 2 93
          NM_001324201.1 NP_001311130.1 2, 3 191
          NM_001324206.1 NP_001311135.1 2, 3 193
          NM_021783.4 NP_068555.1 2, 3 193
          NR_136726.1 2, 3 191
          NR_136727.1 2, 3 193
          XM_011531000.1 XP_011529302.1 2, 3 143
          XM_011531001.1 XP_011529303.1 2, 3 191
          XM_011531002.1 XP_011529304.1 2, 3 143
          XM_011531003.2 XP_011529305.1 2, 3 183
          XM_011531004.1 XP_011529306.1 2, 3 143
          XM_011531006.2 XP_011529308.1 2, 3 183
          XM_011531007.2 XP_011529309.1 2, 3 183
          XM_017029703.1 XP_016885192.1 2, 3 183
          XM_017029704.1 XP_016885193.1 2, 3 143
          XM_017029706.1 XP_016885195.1 2, 3 143
          GenBank mRNA AF298812.1 - 1, 2 83
          AK296264.1 - 2, 3 143
          AK314881.1 - 2, 3 159
          AY152724.1 - 1, 2 83
          AY358735.1 - 1, 2 86
          AY358736.1 - 1, 2 86
          BC034919.1 - 2, 3 140 5434545
          BP249106.1 - 2, 3 159
          DA215848.1 - 2, 3 192
          DA574050.1 - 2, 3 143

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          epidermis development
          tissue development
          ectodermal cell differentiation
          tumor necrosis factor-mediated signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of JNK cascade
          positive regulation of NF-kappaB transcription factor activity
          intrinsic apoptotic signaling pathway by p53 class mediator
          receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0