Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301129.1 | NP_001288058.1 | 5, 6 | 516 | ||
NM_021974.4 | NP_068809.1 | 4, 5 | 418 | |||
NR_125371.1 | 4, 5 | 477 | ||||
GenBank mRNA | AK311831.1 | - | 4, 5 | 382 | ||
BC003582.1 | - | 4, 5 | 396 | 3546712 | ||
CR456546.1 | - | 4, 5 | 308 | |||
CR457021.1 | - | 4, 5 | 293 | |||
DA083933.1 | - | 5, 6 | 480 | |||
DA203006.1 | - | 4, 5 | 441 | |||
DA372532.1 | - | 4, 5 | 418 |