Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130156.2 | NP_001123628.1 | 2 | 334 | ||
NM_001130157.2 | NP_001123629.1 | 3 | 407 | |||
NM_001195432.1 | NP_001182361.1 | 3 | 564 | |||
NM_001195433.1 | NP_001182362.1 | 3 | 407 | |||
NM_001195434.1 | NP_001182363.1 | 3 | 407 | |||
NM_022443.4 | NP_071888.1 | 2 | 290 | |||
XM_005247479.4 | XP_005247536.1 | 2 | 292 | |||
XM_005247480.2 | XP_005247537.1 | 2 | 276 | |||
XM_011512852.2 | XP_011511154.1 | 3 | 365 | |||
XM_011512853.2 | XP_011511155.1 | 3 | 357 | |||
GenBank mRNA | AK056948.1 | - | 3 | 377 | ||
AK096889.1 | - | 3 | 473 | |||
AK297488.1 | - | 2 | 276 | |||
AK315402.1 | - | 2 | 260 | |||
AY848700.1 | - | 3 | 376 | |||
AY848701.1 | - | 3 | 376 | |||
AY848702.1 | - | 2 | 303 | |||
BC007045.1 | - | 2 | 259 | 3996145 | ||
BX647666.1 | - | 2 | 305 | |||
KU178186.1 | - | 2 | 153 | |||
KU178187.1 | - | 2 | 153 | |||
L49054.1 | - | 2 | 261 |