Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543544.1 | XP_011541846.1 | 8 | 1532 | ||
GenBank mRNA | AB109031.1 | - | 8 | 1499 | ||
AB163917.1 | - | 8 | 1499 | |||
AF191545.1 | - | 7 | 1471 | |||
AK074281.1 | - | 8 | 1576 | |||
AK074387.1 | - | 1 | 80 | |||
AK312864.1 | - | 8 | 1512 | |||
AY028805.1 | - | 7 | 1429 | |||
BC065240.1 | - | 7 | 1358 | 6166289 |