Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316329.1 | NP_001303258.1 | 1 | 403 | ||
NM_002739.4 | NP_002730.1 | 1 | 403 | |||
GenBank mRNA | AB209671.1 | - | 1 | 122 | ||
AF345987.1 | - | 1 | 287 | |||
AK296412.1 | - | 1 | 411 | |||
AK296434.1 | - | 1 | 390 | |||
AK302958.1 | - | 1 | 388 | |||
AK312302.1 | - | 1 | 365 | |||
BC047876.1 | - | 1 | 278 | 5764695 | ||
BI757265.1 | - | 1 | 376 | |||
DC318717.1 | - | 1 | 403 | |||
M13977.1 | - | 1 | 287 |