Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMBOX1 SIRNA Assays ›
Gene Symbol:
HMBOX1
Gene Name:
homeobox containing 1
Gene Aliases:
HNF1LA, HOT1, PBHNF
Chromosome Location:
Chr.8: 28890370 - 29064939 on Build GRCh38
UniGene ID:
Hs.563560
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31102
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMBOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135726.2
NP_001129198.1
3
808
NM_001324382.1
NP_001311311.1
3
668
NM_001324383.1
NP_001311312.1
3
808
NM_001324384.1
NP_001311313.1
3
811
NM_001324385.1
NP_001311314.1
3
808
NM_001324386.1
NP_001311315.1
3
808
NM_001324387.1
NP_001311316.1
3
808
NM_001324388.1
NP_001311317.1
3
668
NM_001324389.1
NP_001311318.1
3
808
NM_001324390.1
NP_001311319.1
3
811
NM_001324391.1
NP_001311320.1
3
808
NM_001324392.1
NP_001311321.1
3
811
NM_001324393.1
NP_001311322.1
4
924
NM_001324394.1
NP_001311323.1
3
808
NM_001324395.1
NP_001311324.1
3
808
NM_024567.4
NP_078843.2
4
842
NR_136757.1
3
808
NR_136758.1
3
808
XM_005273635.1
XP_005273692.1
3
784
XM_005273641.4
XP_005273698.1
3
784
XM_006716398.2
XP_006716461.1
3
784
XM_011544642.2
XP_011542944.1
3
784
XM_017013823.1
XP_016869312.1
3
816
XM_017013824.1
XP_016869313.1
3
606
XM_017013825.1
XP_016869314.1
3
784
GenBank mRNA
AK025269.1
-
1
273
AK290683.1
-
3
784
AK295320.1
-
2
447
AK310959.1
-
3
774
AY522342.1
-
3
553
BC009259.2
-
1
226
BC069242.1
-
4
842
5928365
DQ153248.1
-
1
154
DQ269478.1
-
3
553
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA-dependent DNA biosynthetic process
transcription, DNA-templated
positive regulation of telomere maintenance via telomerase
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of telomerase activity
double-stranded telomeric DNA binding
telomerase activity
protein binding
protein complex binding
telomeric DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0