Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31298
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321115.1
NP_001308044.1
23
2756
XM_005252593.2
XP_005252650.1
4
543
XM_006717505.1
XP_006717568.1
21
2251
XM_006717506.1
XP_006717569.1
20
2143
XM_011519663.1
XP_011517965.1
23
2464
XM_011519664.1
XP_011517966.1
23
2464
XM_011519666.1
XP_011517968.1
23
2464
XM_011519667.1
XP_011517969.1
23
2464
XM_011519668.1
XP_011517970.1
22
2374
XM_011519669.1
XP_011517971.1
22
2362
XM_011519670.1
XP_011517972.1
22
2356
XM_011519671.1
XP_011517973.1
23
2464
XM_011519672.1
XP_011517974.1
23
2464
XM_011519673.1
XP_011517975.1
23
2464
XM_011519674.1
XP_011517976.1
23
2464
XM_011519675.1
XP_011517977.1
21
2254
XM_011519676.1
XP_011517978.1
23
2464
XM_011519677.1
XP_011517979.1
22
2242
XM_011519678.1
XP_011517980.1
23
2464
XM_011519679.1
XP_011517981.1
20
1933
XM_011519680.2
XP_011517982.1
18
1599
XM_011519683.2
XP_011517985.1
12
1273
XM_011519684.2
XP_011517986.1
23
2464
XM_011519685.1
XP_011517987.1
15
1481
XM_011519686.2
XP_011517988.1
23
2464
XM_011519687.1
XP_011517989.1
5
388
XM_017016638.1
XP_016872127.1
23
2464
XM_017016639.1
XP_016872128.1
23
2464
XM_017016640.1
XP_016872129.1
23
2464
XM_017016641.1
XP_016872130.1
18
1893
XM_017016642.1
XP_016872131.1
18
1966
XM_017016643.1
XP_016872132.1
18
1583
XM_017016644.1
XP_016872133.1
23
2464
XM_017016645.1
XP_016872134.1
17
1507
XM_017016646.1
XP_016872135.1
14
1394
XM_017016647.1
XP_016872136.1
23
2464
XM_017016648.1
XP_016872137.1
10
1115
XM_017016649.1
XP_016872138.1
10
1055
XM_017016650.1
XP_016872139.1
23
2464
XM_017016651.1
XP_016872140.1
23
2464
XM_017016652.1
XP_016872141.1
7
518
XM_017016653.1
XP_016872142.1
5
428
GenBank mRNA
AK023093.1
-
4
549
BC125090.1
-
7
718
40118176
BC125091.1
-
3
188
40118177
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0