Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD4 SIRNA Assays ›
Gene Symbol:
ACBD4
Gene Name:
acyl-CoA binding domain containing 4
Gene Aliases:
HMFT0700
Chromosome Location:
Chr.17: 45132600 - 45144181 on Build GRCh38
UniGene ID:
Hs.110298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
31329
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135705.2
NP_001129177.1
5
673
NM_001135706.2
NP_001129178.1
5
673
NM_001135707.2
NP_001129179.1
5
673
NM_001321352.1
NP_001308281.1
7
797
NM_001321353.1
NP_001308282.1
7
794
NM_024722.3
NP_078998.1
4
827
XM_006722085.2
XP_006722148.1
6
1043
XM_011525259.2
XP_011523561.1
6
1043
XM_011525261.2
XP_011523563.1
6
1043
XM_017025084.1
XP_016880573.1
6
1043
XM_017025085.1
XP_016880574.1
7
794
XM_017025086.1
XP_016880575.1
8
943
XM_017025087.1
XP_016880576.1
6
1043
XM_017025088.1
XP_016880577.1
6
1043
XM_017025089.1
XP_016880578.1
6
1043
XM_017025090.1
XP_016880579.1
6
1043
XM_017025091.1
XP_016880580.1
6
1043
XM_017025092.1
XP_016880581.1
7
1168
XM_017025093.1
XP_016880582.1
6
1043
XM_017025094.1
XP_016880583.1
4
787
XM_017025095.1
XP_016880584.1
6
1043
XM_017025096.1
XP_016880585.1
6
1043
XM_017025097.1
XP_016880586.1
6
1043
XM_017025098.1
XP_016880587.1
7
794
XM_017025099.1
XP_016880588.1
6
1043
XM_017025100.1
XP_016880589.1
6
1043
XM_017025101.1
XP_016880590.1
6
1043
GenBank mRNA
AB073387.1
-
5
585
AK023384.1
-
4
787
AK075104.1
-
5
626
AK296167.1
-
5
660
AK316359.1
-
5
673
BC029164.1
-
5
612
5188183
BC041143.1
-
7
766
5769682
KU178779.1
-
4
329
KU178780.1
-
4
329
KU178781.1
-
4
329
KU178782.1
-
4
329
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Process(es)
fatty-acyl-CoA binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jwk2j:80/100.66.76.207:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0