Search Thermo Fisher Scientific
Gene Symbol: UGT2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024743.3 | NP_079019.3 | 1 | 337 | ||
NR_024010.1 | 1 | 337 | ||||
XM_011532247.2 | XP_011530549.1 | 1 | 346 | |||
GenBank mRNA | AK025587.1 | - | 1 | 142 | ||
AK225308.1 | - | 1 | 142 | |||
AY358727.1 | - | 1 | 332 | |||
AY542891.1 | - | 1 | 337 | |||
BC130533.1 | - | 1 | 330 | 40146564 |