Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CEP135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025009.4 | NP_079285.2 | 4, 5 | 777 | ||
XM_005265788.3 | XP_005265845.1 | 4, 5 | 748 | |||
XM_006714055.2 | XP_006714118.1 | 4, 5 | 777 | |||
GenBank mRNA | AK023683.1 | - | 4, 5 | 714 | ||
AK309554.1 | - | 1, 2 | 114 | |||
BC136535.1 | - | 4, 5 | 604 | 9020525 | ||
BC136536.1 | - | 4, 5 | 604 | 9020526 |