Search
Search

PDCD1LG2| Gene Symbol: PDCD1LG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025239.3 | NP_079515.2 | 3 | 464 | ||
| XM_005251600.3 | XP_005251657.1 | 3 | 571 | |||
| GenBank mRNA | AF329193.1 | - | 3 | 449 | ||
| AF344424.1 | - | 3 | 464 | |||
| AY254343.1 | - | 2 | 191 | |||
| AY271901.1 | - | 2 | 191 | |||
| AY271902.1 | - | 2 | 191 | |||
| BC074766.2 | - | 3 | 225 | 30915184 | ||
| BC113678.1 | - | 3 | 227 | 8322730 | ||
| BC113680.1 | - | 3 | 227 | 8322732 | ||
| BC143920.1 | - | 3 | 227 | |||
| BX951459.1 | - | 1 | 101 | |||
| DQ336699.1 | - | 2 | 158 | |||
| EF444806.1 | - | 3 | 311 | |||