Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX11 SIRNA Assays ›
          Gene Symbol:
          DDX11 DDX12P LOC642846
          Gene Name:
          DEAD/H-box helicase 11
          DEAD/H-box helicase 12, pseudogene
          DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11-like
          Gene Aliases:
          CHL1, CHLR1, KRG2, WABS
          CHLR2, DDX12
          -
          Chromosome Location:
          Chr.12: 31073574 - 31104799 on Build GRCh38
          Chromosome Location:
          Chr.12: 9417691 - 9448172 on Build GRCh38
          Chromosome Location:
          Chr.12: 9283657 - 9314088 on Build GRCh38
          UniGene ID:
          Hs.443960
          Hs.447869
          Hs.447869
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 32754

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDX11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257144.1 NP_001244073.1 3 356
          NM_001257145.1 NP_001244074.1 3 283
          NM_004399.2 NP_004390.3 3 478
          NM_030653.3 NP_085911.2 3 478
          NM_152438.1 NP_689651.1 3 478
          XM_005253331.3 XP_005253388.1 3 648
          XM_005253333.3 XP_005253390.1 3 648
          XM_006719050.3 XP_006719113.1 3 648
          XM_011520592.2 XP_011518894.1 3 649
          XM_011520595.2 XP_011518897.1 3 648
          XM_011520597.2 XP_011518899.1 3 648
          XM_017018918.1 XP_016874407.1 3 651
          XM_017018919.1 XP_016874408.1 4 714
          XM_017018920.1 XP_016874409.1 3 652
          XM_017018921.1 XP_016874410.1 3 651
          XM_017018922.1 XP_016874411.1 3 444
          XM_017018923.1 XP_016874412.1 3 337
          XM_017018924.1 XP_016874413.1 3 370
          XM_017018925.1 XP_016874414.1 3 442
          XM_017018926.1 XP_016874415.1 3 652
          XM_017018927.1 XP_016874416.1 3 652
          XM_017018928.1 XP_016874417.1 3 651
          XM_017018929.1 XP_016874418.1 3 652
          XM_017018930.1 XP_016874419.1 3 650
          XM_017018931.1 XP_016874420.1 3 444
          XM_017018932.1 XP_016874421.1 3 337
          XM_017018933.1 XP_016874422.1 3 749
          XM_017018934.1 XP_016874423.1 3 444
          XM_017018938.1 XP_016874427.1 3 650
          XM_017018939.1 XP_016874428.1 3 650
          GenBank mRNA AK297607.1 - 3 463
          AK303139.1 - 3 422
          BC050069.1 - 3 268 5271949
          BC050522.1 - 3 307 6471750
          BC111733.1 - 3 478 40035545
          U33833.1 - 3 405
          U75967.1 - 3 368
          U75968.1 - 3 447
          X99583.1 - 3 439
          Gene Symbol: DDX12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033399.1 3 422
          Gene Symbol: LOC642846
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024374.1 3 471
          GenBank mRNA AK296420.1 - 3 479
          AK303618.1 - 3 249
          AK309820.1 - 3 339
          AK310166.1 - 1 25

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          sister chromatid cohesion
          viral process
          DNA duplex unwinding
          IRE1-mediated unfolded protein response
          cell cycle
          double-stranded DNA binding
          single-stranded DNA binding
          RNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          metal ion binding
          4 iron, 4 sulfur cluster binding
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0