Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDX11 SIRNA Assays ›
Gene Symbol:
DDX11
DDX12P
Gene Name:
DEAD/H-box helicase 11
DEAD/H-box helicase 12, pseudogene
Gene Aliases:
CHL1, CHLR1, KRG2, WABS
CHLR2, DDX12
Chromosome Location:
Chr.12: 31073574 - 31104799 on Build GRCh38
Chromosome Location:
Chr.12: 9417691 - 9448172 on Build GRCh38
UniGene ID:
Hs.443960
Hs.447869
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
32842
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DDX11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257144.1
NP_001244073.1
26
2781
NM_001257145.1
NP_001244074.1
26
2713
NM_004399.2
NP_004390.3
25
2758
NM_030653.3
NP_085911.2
26
2908
NM_152438.1
NP_689651.1
26
2903
XM_005253331.3
XP_005253388.1
26
3073
XM_005253333.3
XP_005253390.1
26
3078
XM_006719050.3
XP_006719113.1
26
3078
XM_011520592.2
XP_011518894.1
26
3074
XM_011520595.2
XP_011518897.1
25
2928
XM_017018918.1
XP_016874407.1
27
3256
XM_017018919.1
XP_016874408.1
28
3319
XM_017018920.1
XP_016874409.1
27
3218
XM_017018921.1
XP_016874410.1
27
3256
XM_017018922.1
XP_016874411.1
27
3049
XM_017018923.1
XP_016874412.1
27
2942
XM_017018924.1
XP_016874413.1
27
2975
XM_017018925.1
XP_016874414.1
27
3047
XM_017018926.1
XP_016874415.1
27
3116
XM_017018927.1
XP_016874416.1
26
3107
XM_017018928.1
XP_016874417.1
27
3261
XM_017018929.1
XP_016874418.1
27
3223
XM_017018930.1
XP_016874419.1
27
3260
XM_017018931.1
XP_016874420.1
26
2874
XM_017018932.1
XP_016874421.1
26
2767
XM_017018933.1
XP_016874422.1
27
3343
XM_017018934.1
XP_016874423.1
25
2795
XM_017018935.1
XP_016874424.1
23
2279
XM_017018936.1
XP_016874425.1
23
3274
XM_017018937.1
XP_016874426.1
23
3279
GenBank mRNA
BC012834.2
-
2
165
BC050069.1
-
26
2698
5271949
BC050522.1
-
26
2732
6471750
BC111733.1
-
26
2908
40035545
U33833.1
-
26
2835
U75967.1
-
26
2798
U75968.1
-
27
3088
U75969.1
-
18
1753
X99583.1
-
25
2719
Gene Symbol: DDX12P
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_033399.1
26
2866
GenBank mRNA
BC065494.1
-
15
2805
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA helicase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
sister chromatid cohesion
viral process
DNA duplex unwinding
IRE1-mediated unfolded protein response
cell cycle
double-stranded DNA binding
single-stranded DNA binding
RNA binding
ATP-dependent DNA helicase activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0