Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030915.3 | NP_112177.2 | 3 | 2861 | ||
XM_017005059.1 | XP_016860548.1 | 3 | 3019 | |||
GenBank mRNA | AF110224.1 | - | 1 | 1631 | ||
AF258555.1 | - | 1 | 1851 | |||
AK023425.1 | - | 1 | 1927 | |||
AK027232.1 | - | 3 | 2654 | |||
AL136703.1 | - | 3 | 2867 | |||
BC109376.1 | - | 3 | 2868 | 6495944 |