Search
Search

SLC41A2| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032148.3 | NP_115524.3 | 1 | 537 | ||
| XM_005269176.1 | XP_005269233.1 | 2 | 743 | |||
| XM_005269178.1 | XP_005269235.1 | 2 | 638 | |||
| XM_005269179.1 | XP_005269236.1 | 2 | 575 | |||
| XM_006719630.2 | XP_006719693.1 | 3 | 898 | |||
| XM_011538807.2 | XP_011537109.1 | 3 | 839 | |||
| XM_011538808.1 | XP_011537110.1 | 3 | 848 | |||
| XM_011538809.1 | XP_011537111.1 | 3 | 795 | |||
| XM_011538810.2 | XP_011537112.1 | 2 | 665 | |||
| XM_011538811.2 | XP_011537113.1 | 2 | 1121 | |||
| XM_017020013.1 | XP_016875502.1 | 2 | 660 | |||
| XM_017020014.1 | XP_016875503.1 | 3 | 898 | |||
| XM_017020015.1 | XP_016875504.1 | 3 | 898 | |||
| GenBank mRNA | AK027838.1 | - | 1 | 462 | ||
| AL136828.1 | - | 1 | 229 | |||
| BC036734.1 | - | 1 | 509 | 5538264 | ||
| BC106871.1 | - | 1 | 454 | 40006660 | ||
| BC106872.1 | - | 1 | 454 | 40006662 | ||