Search
Search

MEGF11| Gene Symbol: MEGF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032445.2 | NP_115821.2 | 8 | 942 | ||
| XM_017022670.1 | XP_016878159.1 | 7 | 1364 | |||
| XM_017022671.1 | XP_016878160.1 | 7 | 1362 | |||
| XM_017022672.1 | XP_016878161.1 | 7 | 1371 | |||
| XM_017022673.1 | XP_016878162.1 | 7 | 1365 | |||
| XM_017022674.1 | XP_016878163.1 | 2 | 1308 | |||
| XM_017022675.1 | XP_016878164.1 | 2 | 1346 | |||
| GenBank mRNA | AB058677.1 | - | 6 | 735 | ||
| AB300051.1 | - | 7 | 801 | |||
| AL834326.1 | - | 1 | 56 | |||
| BC029999.1 | - | 8 | 974 | |||
| BC117419.1 | - | 8 | 862 | 40125970 | ||
| BC126313.1 | - | 8 | 862 | 8992029 | ||