Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABLIM2 SIRNA Assays ›
          Gene Symbol:
          ABLIM2
          Gene Name:
          actin binding LIM protein family member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 7965310 - 8158832 on Build GRCh38
          UniGene ID:
          Hs.233404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 34253

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABLIM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130083.1 NP_001123555.1 21 2055
          NM_001130084.1 NP_001123556.1 20 1953
          NM_001130085.1 NP_001123557.1 18 1836
          NM_001130086.1 NP_001123558.1 18 1797
          NM_001130087.1 NP_001123559.1 17 1713
          NM_001286688.1 NP_001273617.1 15 1317
          NM_032432.4 NP_115808.3 16 1683
          XM_005248014.4 XP_005248071.1 23 2179
          XM_005248017.4 XP_005248074.1 21 2062
          XM_005248018.4 XP_005248075.1 22 2058
          XM_005248019.4 XP_005248076.1 21 2025
          XM_005248020.4 XP_005248077.1 21 2022
          XM_005248021.4 XP_005248078.1 20 1940
          XM_005248022.4 XP_005248079.1 20 1923
          XM_005248023.4 XP_005248080.1 19 1905
          XM_005248024.4 XP_005248081.1 18 1805
          XM_005248027.4 XP_005248084.1 18 1766
          XM_005248028.4 XP_005248085.1 17 1682
          XM_005248029.4 XP_005248086.1 16 1651
          XM_005248030.4 XP_005248087.1 16 1648
          XM_006713923.3 XP_006713986.1 21 2059
          XM_006713924.3 XP_006713987.1 22 2055
          XM_006713925.3 XP_006713988.1 20 1939
          XM_006713927.3 XP_006713990.1 17 1682
          XM_011513586.2 XP_011511888.1 22 2173
          XM_017008706.1 XP_016864195.1 23 2177
          XM_017008707.1 XP_016864196.1 21 2072
          XM_017008708.1 XP_016864197.1 20 2028
          XM_017008709.1 XP_016864198.1 20 1960
          XM_017008710.1 XP_016864199.1 21 1956
          XM_017008711.1 XP_016864200.1 19 1929
          XM_017008712.1 XP_016864201.1 19 1926
          XM_017008713.1 XP_016864202.1 19 1907
          XM_017008714.1 XP_016864203.1 19 1919
          XM_017008715.1 XP_016864204.1 18 1851
          XM_017008716.1 XP_016864205.1 19 1841
          XM_017008717.1 XP_016864206.1 19 1838
          XM_017008718.1 XP_016864207.1 19 1850
          XM_017008719.1 XP_016864208.1 18 1808
          XM_017008720.1 XP_016864209.1 19 1798
          XM_017008721.1 XP_016864210.1 18 1768
          XM_017008722.1 XP_016864211.1 17 1751
          XM_017008724.1 XP_016864213.1 17 1697
          XM_017008726.1 XP_016864215.1 17 1515
          XM_017008727.1 XP_016864216.1 15 1398
          GenBank mRNA AB058711.1 - 17 1595
          AJ748600.1 - 18 1758
          AJ748601.1 - 20 1875
          AK094754.1 - 17 1692
          AK094798.1 - 15 1287
          AK097003.1 - 2 257
          AL834195.1 - 7 1050
          BC122567.1 - 16 1634 40085570
          DQ413176.1 - 21 1944
          DQ413177.1 - 17 1654

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          actin cytoskeleton organization
          actin binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0