Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032775.3 | NP_116164.2 | 6 | 1536 | ||
NR_033825.1 | 5 | 1370 | ||||
XM_017029018.1 | XP_016884507.1 | 5 | 1704 | |||
XM_017029019.1 | XP_016884508.1 | 6 | 1832 | |||
XM_017029020.1 | XP_016884509.1 | 7 | 3011 | |||
XM_017029021.1 | XP_016884510.1 | 6 | 1465 | |||
XM_017029022.1 | XP_016884511.1 | 5 | 1636 | |||
XM_017029023.1 | XP_016884512.1 | 5 | 4096 | |||
XM_017029024.1 | XP_016884513.1 | 5 | 1668 | |||
XM_017029025.1 | XP_016884514.1 | 5 | 1184 | |||
GenBank mRNA | AK024369.1 | - | 1 | 2917 | ||
AK027266.1 | - | 6 | 1475 | |||
AK222850.1 | - | 6 | 1442 | |||
AK290669.1 | - | 6 | 1448 | |||
AK294682.1 | - | 5 | 1282 | |||
BC015923.1 | - | 6 | 1488 | 4562420 | ||
CR456352.1 | - | 5 | 1413 |