Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CORO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032854.3 | NP_116243.2 | 6 | 793 | ||
XM_005258047.3 | XP_005258104.1 | 7 | 1075 | |||
XM_005258048.4 | XP_005258105.2 | 7 | 1012 | |||
XM_005258056.4 | XP_005258113.1 | 2 | 892 | |||
XM_011525385.2 | XP_011523687.1 | 7 | 1012 | |||
XM_011525387.2 | XP_011523689.1 | 6 | 889 | |||
XM_011525388.2 | XP_011523690.1 | 7 | 1012 | |||
XM_011525390.2 | XP_011523692.1 | 3 | 755 | |||
XM_011525391.2 | XP_011523693.1 | 3 | 733 | |||
XM_017025235.1 | XP_016880724.1 | 6 | 956 | |||
GenBank mRNA | AF447884.1 | - | 3 | 777 | ||
AF447885.1 | - | 1 | 463 | |||
AK027777.1 | - | 1 | 465 | |||
AK092430.1 | - | 3 | 777 | |||
AK094683.1 | - | 2 | 1615 | |||
AL833952.1 | - | 4 | 2184 | |||
AY545070.1 | - | 1 | 233 | |||
BC048209.1 | - | 3 | 207 | |||
BC064514.1 | - | 3 | 207 | 3678185 |