Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35100
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
129
23704
NM_182961.3
NP_892006.3
130
24082
XM_006715407.1
XP_006715470.1
130
23995
XM_006715408.2
XP_006715471.1
130
23987
XM_006715409.1
XP_006715472.1
129
23974
XM_006715410.2
XP_006715473.1
130
23999
XM_006715411.1
XP_006715474.1
129
23944
XM_006715412.2
XP_006715475.1
130
23984
XM_006715413.2
XP_006715476.1
130
23999
XM_006715414.1
XP_006715477.1
128
23923
XM_006715415.2
XP_006715478.1
130
23999
XM_006715416.2
XP_006715479.1
130
23984
XM_006715417.2
XP_006715480.1
129
23858
XM_006715420.2
XP_006715483.1
129
23846
XM_006715421.2
XP_006715484.1
129
23843
XM_006715422.1
XP_006715485.1
129
23836
XM_006715423.2
XP_006715486.1
130
23999
XM_006715424.2
XP_006715487.1
130
23999
XM_006715425.2
XP_006715488.1
130
23999
XM_011535641.2
XP_011533943.1
130
23996
XM_011535642.2
XP_011533944.1
130
23984
XM_011535643.1
XP_011533945.1
129
23830
XM_011535644.1
XP_011533946.1
116
22046
XM_011535645.2
XP_011533947.1
112
21439
XM_017010608.1
XP_016866097.1
131
24114
XM_017010609.1
XP_016866098.1
130
23921
XM_017010610.1
XP_016866099.1
129
23900
XM_017010611.1
XP_016866100.1
129
23703
XM_017010612.1
XP_016866101.1
128
23627
XM_017010613.1
XP_016866102.1
130
23996
XM_017010614.1
XP_016866103.1
129
23843
XM_017010615.1
XP_016866104.1
129
23843
XM_017010616.1
XP_016866105.1
130
23999
XM_017010617.1
XP_016866106.1
130
23996
XM_017010618.1
XP_016866107.1
130
23984
XM_017010619.1
XP_016866108.1
116
22078
GenBank mRNA
AB018339.2
-
18
2719
AF444779.1
-
5
592
AF495910.1
-
130
24082
AF535142.1
-
129
23704
AK122691.1
-
10
1362
AK127076.1
-
14
2059
AK304825.1
-
3
284
AK316101.1
-
3
299
AY061755.1
-
45
7172
AY061756.1
-
2
162
AY184206.1
-
43
7067
BC090927.1
-
3
344
BX537837.1
-
12
1660
BX647837.1
-
15
2254
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0