Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT3A SIRNA Assays ›
          Gene Symbol:
          WNT3A
          Gene Name:
          Wnt family member 3A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 228007022 - 228067113 on Build GRCh38
          UniGene ID:
          Hs.336930
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 35303

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033131.3 NP_149122.1 4 2499
          GenBank mRNA AK056278.1 - 4 2470

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          positive regulation of cytokine production
          positive regulation of protein phosphorylation
          heart looping
          positive regulation of receptor internalization
          negative regulation of heart induction by canonical Wnt signaling pathway
          axon guidance
          synapse assembly
          positive regulation of cell proliferation
          COP9 signalosome assembly
          negative regulation of neuron projection development
          Wnt signaling pathway
          spinal cord association neuron differentiation
          hippocampus development
          cell proliferation in forebrain
          Wnt signaling pathway involved in forebrain neuroblast division
          dorsal/ventral neural tube patterning
          hemopoiesis
          neuron differentiation
          extracellular matrix organization
          mammary gland development
          positive regulation of B cell proliferation
          positive regulation of protein binding
          positive regulation of peptidyl-serine phosphorylation
          cell proliferation in midbrain
          cellular protein localization
          positive regulation of catenin import into nucleus
          skeletal muscle cell differentiation
          post-anal tail morphogenesis
          synaptic vesicle recycling
          inner ear morphogenesis
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of mesodermal cell fate specification
          paraxial mesodermal cell fate commitment
          positive regulation of skeletal muscle tissue development
          positive regulation of collateral sprouting in absence of injury
          negative regulation of axon extension involved in axon guidance
          negative regulation of neurogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          palate development
          canonical Wnt signaling pathway
          positive regulation of protein tyrosine kinase activity
          positive regulation of dermatome development
          canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
          regulation of microtubule cytoskeleton organization
          platelet aggregation
          cellular response to retinoic acid
          axis elongation involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          positive regulation of protein localization to plasma membrane
          negative regulation of dopaminergic neuron differentiation
          beta-catenin destruction complex disassembly
          Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          positive regulation of cell-cell adhesion mediated by cadherin
          positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
          positive regulation of neural precursor cell proliferation
          positive regulation of hepatocyte proliferation
          positive regulation of transcription regulatory region DNA binding
          positive regulation of cardiac muscle cell differentiation
          transcription coactivator activity
          receptor binding
          frizzled binding
          frizzled-2 binding
          protein binding
          protein domain specific binding
          co-receptor binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0