Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LARP4 SIRNA Assays ›
          Gene Symbol:
          LARP4
          Gene Name:
          La ribonucleoprotein domain family member 4
          Gene Aliases:
          PP13296
          Chromosome Location:
          Chr.12: 50400809 - 50480005 on Build GRCh38
          UniGene ID:
          Hs.26613
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 35517

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LARP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170803.1 NP_001164274.1 3 460
          NM_001170804.1 NP_001164275.1 3 460
          NM_001170808.1 NP_001164279.1 3 460
          NM_052879.4 NP_443111.4 3 460
          NM_199188.2 NP_954658.2 3 457
          NM_199190.2 NP_954660.1 3 460
          NR_033200.1 4 509
          XM_005268613.2 XP_005268670.1 4 509
          XM_011537828.2 XP_011536130.1 4 564
          XM_011537829.2 XP_011536131.1 4 545
          XM_011537830.2 XP_011536132.1 3 392
          XM_011537831.2 XP_011536133.1 3 456
          XM_011537833.2 XP_011536135.1 4 564
          XM_011537834.2 XP_011536136.1 4 565
          XM_011537835.2 XP_011536137.1 4 545
          XM_011537836.2 XP_011536138.1 4 546
          XM_011537838.2 XP_011536140.1 3 304
          XM_011537839.2 XP_011536141.1 4 448
          XM_011537840.2 XP_011536142.1 4 501
          XM_011537841.2 XP_011536143.1 3 354
          XM_011537843.2 XP_011536145.1 4 546
          XM_011537844.2 XP_011536146.1 4 565
          XM_017018750.1 XP_016874239.1 3 386
          XM_017018751.1 XP_016874240.1 4 325
          XM_017018752.1 XP_016874241.1 3 308
          XM_017018753.1 XP_016874242.1 3 341
          XM_017018754.1 XP_016874243.1 4 491
          XM_017018755.1 XP_016874244.1 3 371
          XM_017018756.1 XP_016874245.1 4 423
          XM_017018757.1 XP_016874246.1 5 593
          XM_017018758.1 XP_016874247.1 4 529
          XM_017018759.1 XP_016874248.1 3 354
          XM_017018760.1 XP_016874249.1 3 460
          XM_017018761.1 XP_016874250.1 3 453
          XM_017018762.1 XP_016874251.1 3 453
          GenBank mRNA AF370416.1 - 3 338
          AK055521.1 - 3 302
          AK125113.1 - 2 250
          AK291746.1 - 3 329
          AL832198.1 - 3 403
          AL833272.1 - 3 370
          AY004310.1 - 4 364
          BC022377.1 - 3 362
          BC063479.1 - 3 361 5219803
          BX647457.1 - 3 284
          CR936626.1 - 3 378
          DB061954.1 - 4 509

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          regulation of cell morphogenesis
          nucleotide binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9wt54:80/100.66.77.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0