Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35602
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
13
1470
NM_001170808.1
NP_001164279.1
14
1683
NM_052879.4
NP_443111.4
15
1896
NM_199188.2
NP_954658.2
15
1893
NM_199190.2
NP_954660.1
14
1683
XM_005268613.2
XP_005268670.1
16
1945
XM_011537828.2
XP_011536130.1
16
2000
XM_011537829.2
XP_011536131.1
16
1981
XM_011537830.2
XP_011536132.1
15
1828
XM_011537831.2
XP_011536133.1
15
1892
XM_011537833.2
XP_011536135.1
15
1787
XM_011537834.2
XP_011536136.1
15
1788
XM_011537835.2
XP_011536137.1
15
1768
XM_011537836.2
XP_011536138.1
15
1769
XM_011537838.2
XP_011536140.1
15
1740
XM_011537839.2
XP_011536141.1
16
1884
XM_011537840.2
XP_011536142.1
16
1937
XM_011537841.2
XP_011536143.1
15
1790
XM_011537843.2
XP_011536145.1
14
1556
XM_011537844.2
XP_011536146.1
15
1879
XM_017018750.1
XP_016874239.1
14
1609
XM_017018751.1
XP_016874240.1
16
1761
XM_017018752.1
XP_016874241.1
15
1744
XM_017018753.1
XP_016874242.1
15
1777
XM_017018754.1
XP_016874243.1
16
1927
XM_017018755.1
XP_016874244.1
14
1594
XM_017018756.1
XP_016874245.1
15
1646
XM_017018757.1
XP_016874246.1
16
1816
XM_017018758.1
XP_016874247.1
15
1752
XM_017018759.1
XP_016874248.1
14
1577
XM_017018760.1
XP_016874249.1
14
1774
XM_017018761.1
XP_016874250.1
13
1554
XM_017018762.1
XP_016874251.1
13
1554
GenBank mRNA
AF370416.1
-
15
1774
AK055521.1
-
13
1444
AK125113.1
-
14
1686
AK291746.1
-
15
1765
AL832198.1
-
14
1624
AL833272.1
-
15
1806
BC022377.1
-
13
1370
BC032444.2
-
2
151
BX647457.1
-
14
1507
CR936626.1
-
15
1814
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0