Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GIT2 SIRNA Assays ›
Gene Symbol:
GIT2
Gene Name:
GIT ArfGAP 2
Gene Aliases:
CAT-2, CAT2, PKL
Chromosome Location:
Chr.12: 109929792 - 109996389 on Build GRCh38
UniGene ID:
Hs.434996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35874
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GIT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135213.1
NP_001128685.1
7, 8
883
NM_001135214.1
NP_001128686.1
7, 8
883
NM_014776.3
NP_055591.2
7, 8
883
NM_057169.3
NP_476510.1
7, 8
883
NM_057170.3
NP_476511.1
7, 8
883
NM_139201.2
NP_631940.2
7, 8
883
XM_005253997.3
XP_005254054.1
7, 8
878
XM_005254000.3
XP_005254057.1
7, 8
878
XM_006719707.3
XP_006719770.1
7, 8
878
XM_006719708.3
XP_006719771.1
7, 8
878
XM_006719709.3
XP_006719772.1
7, 8
878
XM_006719712.3
XP_006719775.1
7, 8
878
XM_006719713.3
XP_006719776.1
7, 8
875
XM_006719714.3
XP_006719777.1
7, 8
878
XM_017020258.1
XP_016875747.1
7, 8
878
XM_017020259.1
XP_016875748.1
7, 8
875
XM_017020260.1
XP_016875749.1
7, 8
875
XM_017020261.1
XP_016875750.1
7, 8
873
GenBank mRNA
AB451294.1
-
7, 8
718
AB451428.1
-
7, 8
718
AF124491.1
-
7, 8
718
AK222736.1
-
7, 8
790
AK303198.1
-
6, 7
584
AK304240.1
-
7, 8
867
AK308304.1
-
7, 8
798
BC001379.2
-
7, 8
783
3051069
BC014223.2
-
6, 7
634
BT007312.1
-
7, 8
718
CR533568.1
-
7, 8
718
D63482.2
-
7, 8
789
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00207498_m1
Gene Ontology Categories:
Function(s)
Process(es)
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
behavioral response to pain
GTPase activator activity
protein binding
protein complex binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0