Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
SLC36A1
Gene Name:
solute carrier family 36 member 1
Gene Aliases:
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location:
Chr.5: 151411344 - 151556318 on Build GRCh38
UniGene ID:
Hs.269004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36091
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC36A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308150.1
NP_001295079.1
4, 5
554
NM_001308151.1
NP_001295080.1
4, 5
524
NM_078483.3
NP_510968.2
4, 5
554
XM_005268386.1
XP_005268443.1
4, 5
524
XM_006714759.3
XP_006714822.1
4, 5
603
XM_011537580.2
XP_011535882.1
4, 5
1244
XM_011537581.1
XP_011535883.1
5, 6
672
XM_011537583.2
XP_011535885.1
5, 6
5109
XM_011537584.2
XP_011535886.1
4, 5
763
XM_011537585.1
XP_011535887.1
5, 6
619
XM_011537586.2
XP_011535888.1
4, 5
1367
XM_011537587.2
XP_011535889.1
4, 5
623
XM_011537588.2
XP_011535890.1
4, 5
467
XM_011537589.2
XP_011535891.1
4, 5
1375
XM_011537590.1
XP_011535892.1
4, 5
382
XM_011537591.1
XP_011535893.1
4, 5
357
XM_011537592.2
XP_011535894.1
4, 5
361
XM_011537593.2
XP_011535895.1
5, 6
617
XM_011537594.1
XP_011535896.1
6, 7
731
XM_011537595.2
XP_011535897.2
4, 5
1250
XM_011537596.2
XP_011535898.2
3, 4
336
XM_017009216.1
XP_016864705.1
4, 5
450
XM_017009217.1
XP_016864706.1
4, 5
554
XM_017009218.1
XP_016864707.1
3, 4
336
XM_017009219.1
XP_016864708.1
4, 5
604
XM_017009220.1
XP_016864709.1
4, 5
524
GenBank mRNA
AF516142.1
-
3, 4
346
AK057340.1
-
4, 5
524
AK309323.1
-
4, 5
526
AY162213.1
-
4, 5
459
AY227111.1
-
5, 6
474
AY227113.1
-
4, 5
430
BC136437.1
-
4, 5
425
9020426
BC136438.1
-
4, 5
425
9020427
BM787446.1
-
4, 5
557
BX537963.1
-
4, 5
551
GU296670.1
-
3, 4
331
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7njqw:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0