Search
Search

GTF3A| Gene Symbol: GTF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002097.2 | NP_002088.2 | 1 | 315 | ||
| GenBank mRNA | BC143711.1 | - | 1 | 381 | ||
| BP254755.1 | - | 1 | 296 | |||
| D32257.1 | - | 1 | 314 | |||
| U14134.1 | - | 1 | 115 | |||
| U20272.1 | - | 1 | 115 | |||