Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGO1 SIRNA Assays ›
Gene Symbol:
SGO1
Gene Name:
shugoshin 1
Gene Aliases:
CAID, NY-BR-85, SGO, SGOL1
Chromosome Location:
Chr.3: 20160593 - 20188143 on Build GRCh38
UniGene ID:
Hs.105153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36743
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012409.3
NP_001012409.1
2
267
NM_001012410.4
NP_001012410.1
2
267
NM_001012411.3
NP_001012411.1
2
267
NM_001012412.4
NP_001012412.1
2
267
NM_001012413.3
NP_001012413.1
2
267
NM_001199251.2
NP_001186180.1
2
378
NM_001199252.2
NP_001186181.1
2
378
NM_001199253.2
NP_001186182.1
2
378
NM_001199254.2
NP_001186183.1
2
378
NM_001199255.2
NP_001186184.1
2
378
NM_001199256.2
NP_001186185.1
2
378
NM_001199257.2
NP_001186186.1
2
378
NM_138484.4
NP_612493.1
2
267
NR_131179.1
2
267
NR_131180.1
2
267
XM_011533373.2
XP_011531675.1
2
351
XM_011533375.2
XP_011531677.1
2
720
XM_011533376.2
XP_011531678.1
2
575
XM_011533377.2
XP_011531679.1
2
1555
GenBank mRNA
AB187577.1
-
2
225
AB187578.1
-
2
225
AB187579.1
-
2
225
AB187580.1
-
2
225
AB187581.1
-
2
225
AB187582.1
-
2
225
AB190994.1
-
2
115
AB193056.1
-
2
272
AB193057.1
-
2
161
AB193058.1
-
2
161
AB193059.1
-
2
272
AB193060.1
-
2
272
AB193061.1
-
2
161
AB193062.1
-
2
272
AB193063.1
-
2
161
AB193064.1
-
2
272
AB193065.1
-
2
272
AB193066.1
-
2
161
AB451340.1
-
1
85
AB451489.1
-
1
85
AB567656.1
-
2
225
AB567657.1
-
2
225
AK308095.1
-
2
366
BC001339.2
-
26
BC017867.1
-
2
225
4685140
BG530826.1
-
2
239
BX648516.1
-
2
352
DB163747.1
-
2
377
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
attachment of spindle microtubules to kinetochore
centriole-centriole cohesion
meiotic chromosome segregation
cell division
protein binding
kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0