Search
Search

KIF23| Gene Symbol: KIF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281301.1 | NP_001268230.1 | 3, 4 | 435 | ||
| NM_004856.6 | NP_004847.2 | 3, 4 | 435 | |||
| NM_138555.3 | NP_612565.1 | 3, 4 | 435 | |||
| XM_005254796.2 | XP_005254853.1 | 3, 4 | 384 | |||
| XM_005254797.2 | XP_005254854.1 | 3, 4 | 384 | |||
| XM_005254798.2 | XP_005254855.1 | 3, 4 | 384 | |||
| XM_005254799.2 | XP_005254856.1 | 3, 4 | 384 | |||
| XM_011522238.1 | XP_011520540.1 | 3, 4 | 356 | |||
| XM_011522239.2 | XP_011520541.1 | 3, 4 | 286 | |||
| XM_017022733.1 | XP_016878222.1 | 3, 4 | 384 | |||
| GenBank mRNA | AK308322.1 | - | 3, 4 | 357 | ||
| AK310402.1 | - | 3, 4 | 332 | |||
| AU133373.1 | - | 3, 4 | 332 | |||
| BC017705.2 | - | 2, 3 | 203 | |||
| BC051826.1 | - | 3, 4 | 377 | 6500787 | ||
| DB445953.1 | - | 3, 4 | 434 | |||
| DR000298.1 | - | 3, 4 | 399 | |||
| X67155.2 | - | 2, 3 | 275 | |||