Search
Search

KIF23| Gene Symbol: KIF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281301.1 | NP_001268230.1 | 8, 9 | 1060 | ||
| NM_004856.6 | NP_004847.2 | 9, 10 | 1197 | |||
| NM_138555.3 | NP_612565.1 | 9, 10 | 1197 | |||
| XM_005254796.2 | XP_005254853.1 | 10, 11 | 1188 | |||
| XM_005254797.2 | XP_005254854.1 | 10, 11 | 1188 | |||
| XM_005254798.2 | XP_005254855.1 | 10, 11 | 1188 | |||
| XM_005254799.2 | XP_005254856.1 | 10, 11 | 1188 | |||
| XM_011522238.1 | XP_011520540.1 | 9, 10 | 1023 | |||
| XM_011522239.2 | XP_011520541.1 | 9, 10 | 1048 | |||
| XM_017022733.1 | XP_016878222.1 | 10, 11 | 1188 | |||
| GenBank mRNA | AK303874.1 | - | 5, 6 | 591 | ||
| AK308322.1 | - | 8, 9 | 982 | |||
| AK308361.1 | - | 3, 4 | 406 | |||
| BC017705.2 | - | 8, 9 | 965 | |||
| BC051826.1 | - | 9, 10 | 1139 | 6500787 | ||
| X67155.2 | - | 8, 9 | 1037 | |||